N,2,2,6,6-Pentamethylpiperidin-4-amine 98%

Lieferant: Apollo Scientific

OR904295-1G OR904295-5G
APOSOR904295-1GEA 87 EUR
APOSOR904295-1G APOSOR904295-5G
N,2,2,6,6-Pentamethylpiperidin-4-amine 98%
N,2,2,6,6-Pentamethylpiperidin-4-amine
Formel: C₁₀H₂₂N₂
Molecular Weight: 170,295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08059893
CAS-Nummer: 62995-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR