(R)-3-(1-Aminoethyl)benzonitrile hydrochloride 95%

Lieferant: Apollo Scientific

OR904431-1G OR904431-250MG
APOSOR904431-1GEA 387 EUR
APOSOR904431-1G APOSOR904431-250MG
(R)-3-(1-Aminoethyl)benzonitrile hydrochloride 95%
(R)-3-(1-Aminoethyl)benzonitrile hydrochloride
Formel: C₉H₁₀N₂·ClH
Molecular Weight: 182.65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11101227
CAS-Nummer: 1286693-23-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR