2-tert-Butylpyrrolidine hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
2-(tert-Butyl)pyrrolidine hydrochloride
OR904587-100MG
OR904587-1G
OR904587-250MG
APOSOR904587-100MGEA
209
EUR
APOSOR904587-100MG
APOSOR904587-1G
APOSOR904587-250MG
2-tert-Butylpyrrolidine hydrochloride 95%
2-tert-Butylpyrrolidine hydrochloride
Formel:
C₈H₁₇N·ClH Molecular Weight: 163,688 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07357529 CAS-Nummer: 558478-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...