2-Furan-2-yl-azepane 95%
Lieferant: Apollo Scientific
OR904611-1G
OR904611-250MG
APOSOR904611-1GEA
632
EUR
APOSOR904611-1G
APOSOR904611-250MG
2-Furan-2-yl-azepane 95%
2-Furan-2-yl-azepane
Formel:
C₁₀H₁₅NO Molecular Weight: 165,232 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02663713 CAS-Nummer: 383129-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...