7-Bromo[1,2,4]triazolo[4,3-a]pyridine 98%

Lieferant: Apollo Scientific

Synonyme: 7-Bromo-[1,2,4]triazolo[4,3-a]pyridine

OR904778-1G
APOSOR904778-1GEA 74.1 EUR
APOSOR904778-1G
7-Bromo[1,2,4]triazolo[4,3-a]pyridine 98%
7-Bromo[1,2,4]triazolo[4,3-a]pyridine
Formel: C₆H₄BrN₃
Molecular Weight: 198,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09994418
CAS-Nummer: 832735-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR