4-Methyl-1-(pyrrolidin-2-ylmethyl)piperidine 95%
Lieferant: Apollo Scientific
OR904846-100MG
OR904846-1G
OR904846-250MG
APOSOR904846-100MGEA
190
EUR
APOSOR904846-100MG
APOSOR904846-1G
APOSOR904846-250MG
4-Methyl-1-(pyrrolidin-2-ylmethyl)piperidine 95%
4-Methyl-1-(pyrrolidin-2-ylmethyl)piperidine
Formel:
C₁₁H₂₂N₂ Molecular Weight: 182,306 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06653508 CAS-Nummer: 881041-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...