2-Butoxypyridine-3-boronic acid 98%

Lieferant: Apollo Scientific

OR904977-1G OR904977-5G
APOSOR904977-1GEA 219 EUR
APOSOR904977-1G APOSOR904977-5G
2-Butoxypyridine-3-boronic acid 98%
2-Butoxypyridine-3-boronic acid
Formel: C₉H₁₄BNO₃
Molecular Weight: 195,023 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20441822
CAS-Nummer: 1987879-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR