2-Isopropoxynitrobenzene 98%

Lieferant: Apollo Scientific

OR905066-1G OR905066-5G
APOSOR905066-1GEA 102 EUR
APOSOR905066-1G APOSOR905066-5G
2-Isopropoxynitrobenzene 98%
2-Isopropoxynitrobenzene
Formel: C₉H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01657560
CAS-Nummer: 38753-50-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR