1-(2-Pyrrolidinylmethyl)azepane 98%

Lieferant: Apollo Scientific

OR905308-1G OR905308-250MG
APOSOR905308-1GEA 348 EUR
APOSOR905308-1G APOSOR905308-250MG
1-(2-Pyrrolidinylmethyl)azepane 98%
1-(2-Pyrrolidinylmethyl)azepane
Formel: C₁₁H₂₂N₂
Molecular Weight: 182,306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06653331
CAS-Nummer: 881040-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR