2-[(Methylsulphanyl)methyl]piperidine hydrochloride 95%

Lieferant: Apollo Scientific

OR905512-100MG OR905512-1G OR905512-250MG
APOSOR905512-100MGEA 190 EUR
APOSOR905512-100MG APOSOR905512-1G APOSOR905512-250MG
2-[(Methylsulphanyl)methyl]piperidine hydrochloride 95%
2-[(Methylsulphanyl)methyl]piperidine hydrochloride
Formel: C₇H₁₅NS·ClH
Molecular Weight: 181,727 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07357622
CAS-Nummer: 1177350-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR