Ethyl (2s)-2-(2-formyl-1H-pyrrol-1-yl)propanoate 95%

Lieferant: Apollo Scientific

OR905661-1G OR905661-250MG
APOSOR905661-1GEA 348 EUR
APOSOR905661-1G APOSOR905661-250MG
Ethyl (2s)-2-(2-formyl-1H-pyrrol-1-yl)propanoate 95%
Ethyl (2s)-2-(2-formyl-1H-pyrrol-1-yl)propanoate
Formel: C₁₀H₁₃NO₃
Molecular Weight: 195,215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832433
CAS-Nummer: 935765-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR