3-[(Methoxymethoxy)methyl]phenylboronic acid 96%

Lieferant: Apollo Scientific

OR905810-1G OR905810-5G
APOSOR905810-1GEA 183 EUR
APOSOR905810-1G APOSOR905810-5G
3-[(Methoxymethoxy)methyl]phenylboronic acid 96%
3-[(Methoxymethoxy)methyl]phenylboronic acid
Formel: C₉H₁₃BO₄
Molecular Weight: 196.008 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205753
CAS-Nummer: 2096342-14-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR