2-Neopentylpiperidine 95%

Lieferant: Apollo Scientific

OR905847-100MG OR905847-1G OR905847-250MG
APOSOR905847-100MGEA 190 EUR
APOSOR905847-100MG APOSOR905847-1G APOSOR905847-250MG
2-Neopentylpiperidine 95%
2-Neopentylpiperidine
Formel: C₁₀H₂₁N
Lagertemperatur: Raumtemperatur
MDL: MFCD02663605
CAS-Nummer: 383128-30-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR