1,4-Dichloro-2-isocyano-benzene 95%

Lieferant: Apollo Scientific

OR905936-100MG OR905936-1G OR905936-250MG
APOSOR905936-100MGEA 190 EUR
APOSOR905936-100MG APOSOR905936-1G APOSOR905936-250MG
1,4-Dichloro-2-isocyano-benzene 95%
1,4-Dichloro-2-isocyano-benzene
Formel: C₇H₃Cl₂N
Molecular Weight: 172.011 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664598
CAS-Nummer: 245539-10-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR