3-(Cyclopropylcarbonyl)phenylboronic acid 95%

Lieferant: Apollo Scientific

OR906064-1G OR906064-5G
APOSOR906064-1GEA 220 EUR
APOSOR906064-1G APOSOR906064-5G
3-(Cyclopropylcarbonyl)phenylboronic acid 95%
3-(Cyclopropylcarbonyl)phenylboronic acid
Formel: C₁₀H₁₁BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22383671
CAS-Nummer: 2377609-66-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR