(5-Methoxypyridin-2-yl)methanol 95%
Lieferant: Apollo Scientific
Synonyme:
(5-Methoxypyridine-2-yl)methanol
OR906110-100MG
OR906110-1G
OR906110-250MG
APOSOR906110-100MGEA
56.7
EUR
APOSOR906110-100MG
APOSOR906110-1G
APOSOR906110-250MG
(5-Methoxypyridin-2-yl)methanol 95%
(5-Methoxypyridin-2-yl)methanol
Formel:
C₇H₉NO₂ Molecular Weight: 139.152 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08235020 CAS-Nummer: 127978-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...