2-Amino-N-methyl-DL-propanamide 98%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-N-methylpropanamide

OR906370-1G OR906370-5G
APOSOR906370-1GEA 114 EUR
APOSOR906370-1G APOSOR906370-5G
2-Amino-N-methyl-DL-propanamide 98%
2-Amino-N-methyl-DL-propanamide
Formel: C₄H₁₀N₂O
Molecular Weight: 102,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09927582
CAS-Nummer: 32012-12-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR