2-(4-Chlorobenzyl)pyrrolidine 95%

Lieferant: Apollo Scientific

OR906442-1G OR906442-5G
APOSOR906442-1GEA 364 EUR
APOSOR906442-1G APOSOR906442-5G
2-(4-Chlorobenzyl)pyrrolidine 95%
2-(4-Chlorobenzyl)pyrrolidine
Formel: C₁₁H₁₄ClN
Molecular Weight: 195,689 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02663506
CAS-Nummer: 66162-29-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR