Methyl D-homoalaninate hydrochloride 98%

Lieferant: Apollo Scientific

Synonyme: H-D-2-Abu-OMe·HCl, (R)-Methyl 2-aminobutanoate hydrochloride

OR906452-100G OR906452-25G
APOSOR906452-100GEA 241 EUR
APOSOR906452-100G APOSOR906452-25G
Methyl D-homoalaninate hydrochloride 98%
Methyl D-homoalaninate hydrochloride
Formel: C₅H₁₁NO₂·ClH
Molecular Weight: 153,607 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10565758
CAS-Nummer: 85774-09-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR