2-Isopropylpiperidine hydrochloride 95%

Lieferant: Apollo Scientific

OR906549-100MG OR906549-1G OR906549-250MG
APOSOR906549-100MGEA 190 EUR
APOSOR906549-100MG APOSOR906549-1G APOSOR906549-250MG
2-Isopropylpiperidine hydrochloride 95%
2-Isopropylpiperidine hydrochloride
Formel: C₈H₁₇N·ClH
Molecular Weight: 163,688 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07357628
CAS-Nummer: 1177339-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR