N-Propylpiperidine-4-carboxamide 96%

Lieferant: Apollo Scientific

OR906944-1G OR906944-5G
APOSOR906944-1GEA 284 EUR
APOSOR906944-1G APOSOR906944-5G
N-Propylpiperidine-4-carboxamide 96%
N-Propylpiperidine-4-carboxamide
Formel: C₉H₁₈N₂O
Molecular Weight: 170.252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08444678
CAS-Nummer: 200267-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR