1-Ethyl-1H-pyrazol-5-ylboronic acid 95%
Lieferant: Apollo Scientific
Synonyme:
(1-Ethyl-1H-pyrazol-5-yl)boronic acid
OR907353-5G
APOSOR907353-5GEA
192
EUR
APOSOR907353-5G
1-Ethyl-1H-pyrazol-5-ylboronic acid 95%
1-Ethyl-1H-pyrazol-5-ylboronic acid
Formel:
C₅H₉BN₂O₂ Molecular Weight: 139,948 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419491 CAS-Nummer: 1095080-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...