5-Methyl-2-propoxypyridine-3-boronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (5-Methyl-2-propoxypyridin-3-yl)boronic acid

OR907369-1G OR907369-5G
APOSOR907369-1GEA 139 EUR
APOSOR907369-1G APOSOR907369-5G
5-Methyl-2-propoxypyridine-3-boronic acid 98%
5-Methyl-2-propoxypyridine-3-boronic acid
Formel: C₉H₁₄BNO₃
Molecular Weight: 195.023 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18783119
CAS-Nummer: 2096332-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR