[4-(methoxycarbonyl)-3-methylphenyl]boronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (4-(Methoxycarbonyl)-3-methylphenyl)boronic acid, 3-Methyl-4-(methoxycarbonyl)phenylboronic acid

OR907412-1G OR907412-250MG
APOSOR907412-1GEA 256 EUR
APOSOR907412-1G APOSOR907412-250MG
[4-(methoxycarbonyl)-3-methylphenyl]boronic acid 98%
[4-(methoxycarbonyl)-3-methylphenyl]boronic acid
Formel: C₉H₁₁BO₄
Molecular Weight: 193,992 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18432535
CAS-Nummer: 603122-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR