(2,3,4,5,6-Pentadeuteriophenyl)boronic acid 98%

Lieferant: Apollo Scientific

Synonyme: Phenyl-d5-boronic acid

OR907458-1G OR907458-5G
APOSOR907458-1GEA 168 EUR
APOSOR907458-1G APOSOR907458-5G
(2,3,4,5,6-Pentadeuteriophenyl)boronic acid 98%
(2,3,4,5,6-Pentadeuteriophenyl)boronic acid
Formel: C₆H₂BD₅O₂
Molecular Weight: 126,96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02093711
CAS-Nummer: 215527-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR