Methyl 5-hydroxy-3-methylbenzoate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-hydroxy-5-Methylbenzoate
OR907668-1G
OR907668-250MG
APOSOR907668-1GEA
828
EUR
APOSOR907668-1G
APOSOR907668-250MG
Methyl 5-hydroxy-3-methylbenzoate 95%
Methyl 5-hydroxy-3-methylbenzoate
Formel:
C₉H₁₀O₃ Molecular Weight: 166,174 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11007825 CAS-Nummer: 2615-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...