Methyl 2-chloro-2-phenylacetate 98%

Lieferant: Apollo Scientific

OR907812-25G OR907812-5G
APOSOR907812-25GEA 551 EUR
APOSOR907812-25G APOSOR907812-5G
Methyl 2-chloro-2-phenylacetate 98%
Methyl 2-chloro-2-phenylacetate
Formel: C₉H₉ClO₂
Molecular Weight: 184.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00040996
CAS-Nummer: 7476-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR