Methyl 2,6-dimethylbenzoate 98%

Lieferant: Apollo Scientific

OR907888-1G OR907888-5G
APOSOR907888-1GEA 62.5 EUR
APOSOR907888-1G APOSOR907888-5G
Methyl 2,6-dimethylbenzoate 98%
Methyl 2,6-dimethylbenzoate
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203721
CAS-Nummer: 14920-81-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR