(E)-Ethyl 3-(thiophen-3-yl)acrylate 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-(thiophen-3-yl)acrylate

OR907901-1G OR907901-5G
APOSOR907901-1GEA 62.5 EUR
APOSOR907901-1G APOSOR907901-5G
(E)-Ethyl 3-(thiophen-3-yl)acrylate 98%
(E)-Ethyl 3-(thiophen-3-yl)acrylate
Formel: C₉H₁₀O₂S
Molecular Weight: 182.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07778410
CAS-Nummer: 50266-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR