Ethyl 3-(thiophen-3-yl)propanoate 96%
Lieferant: Apollo Scientific
OR907903-1G
OR907903-5G
APOSOR907903-1GEA
219
EUR
APOSOR907903-1G
APOSOR907903-5G
Ethyl 3-(thiophen-3-yl)propanoate 96%
Ethyl 3-(thiophen-3-yl)propanoate
Formel:
C₉H₁₂O₂S Molecular Weight: 184,255 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18783158 CAS-Nummer: 99198-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...