3-Bromofuran-2-carboxylic acid 95%
Lieferant: Apollo Scientific
OR907958-1G
OR907958-250MG
APOSOR907958-1GEA
166
EUR
APOSOR907958-1G
APOSOR907958-250MG
3-Bromofuran-2-carboxylic acid 95%
3-Bromofuran-2-carboxylic acid
Formel:
C₅H₃BrO₃ Molecular Weight: 190,98 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092761 CAS-Nummer: 14903-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...