3-Chloro-5-methylbenzaldehyde 98%

Lieferant: Apollo Scientific

OR907959-1G OR907959-5G
APOSOR907959-1GEA 31.9 EUR
APOSOR907959-1G APOSOR907959-5G
3-Chloro-5-methylbenzaldehyde 98%
3-Chloro-5-methylbenzaldehyde
Formel: C₈H₇ClO
Molecular Weight: 154.594 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11553565
CAS-Nummer: 103426-20-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR