Methyl 4-carbamoylbenzoate 98%

Lieferant: Apollo Scientific

OR907970-1G OR907970-5G
APOSOR907970-1GEA 154 EUR
APOSOR907970-1G APOSOR907970-5G
Methyl 4-carbamoylbenzoate 98%
Methyl 4-carbamoylbenzoate
Formel: C₉H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00463146
CAS-Nummer: 6757-31-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR