Methyl 1,3-benzodiazole-4-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 1H-benzo[d]imidazole-4-carboxylate

OR908067-1G
APOSOR908067-1GEA 136 EUR
APOSOR908067-1G
Methyl 1,3-benzodiazole-4-carboxylate 98%
Methyl 1,3-benzodiazole-4-carboxylate
Formel: C₉H₈N₂O₂
Molecular Weight: 176,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458706
CAS-Nummer: 37619-25-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR