3-Bromo-4-(hydroxymethyl)benzonitrile 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-4-hydroxymethylbenzonitrile

OR908106-1G OR908106-250MG
APOSOR908106-1GEA 130 EUR
APOSOR908106-1G APOSOR908106-250MG
3-Bromo-4-(hydroxymethyl)benzonitrile 98%
3-Bromo-4-(hydroxymethyl)benzonitrile
Formel: C₈H₆BrNO
Molecular Weight: 212,043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907613
CAS-Nummer: 90110-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR