3,6-Dihydro-2H-pyran-4-carbonitrile 95%
Lieferant: Apollo Scientific
OR908119-1G
APOSOR908119-1GEA
106
EUR
APOSOR908119-1G
3,6-Dihydro-2H-pyran-4-carbonitrile 95%
3,6-Dihydro-2H-pyran-4-carbonitrile
Formel:
C₆H₇NO Molecular Weight: 109,126 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20040386 CAS-Nummer: 105772-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...