Methyl 2-(benzyloxy)acetate 98%
Lieferant: Apollo Scientific
OR908129-5G
APOSOR908129-5GEA
53.8
EUR
APOSOR908129-5G
Methyl 2-(benzyloxy)acetate 98%
Methyl 2-(benzyloxy)acetate
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11174673 CAS-Nummer: 31600-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...