4-Chloro-3-hydroxybenzonitrile 98%

Lieferant: Apollo Scientific

OR908131-1G OR908131-250MG
APOSOR908131-1GEA 104 EUR
APOSOR908131-1G APOSOR908131-250MG
4-Chloro-3-hydroxybenzonitrile 98%
4-Chloro-3-hydroxybenzonitrile
Formel: C₇H₄ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD16999195
CAS-Nummer: 51748-01-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR