2,3-Dihydro-1,4-benzodioxine-5-carbonitrile 98%
Lieferant: Apollo Scientific
Synonyme:
2,3-Dihydrobenzo[b][1,4]dioxine-5-carbonitrile
OR908136-1G
APOSOR908136-1GEA
102
EUR
APOSOR908136-1G
2,3-Dihydro-1,4-benzodioxine-5-carbonitrile 98%
2,3-Dihydro-1,4-benzodioxine-5-carbonitrile
Formel:
C₉H₇NO₂ Molecular Weight: 161,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01116986 CAS-Nummer: 148703-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...