4-Hydroxy-N,N-dimethylbenzamide 98%

Lieferant: Apollo Scientific

OR908142-1G OR908142-5G
APOSOR908142-1GEA 75.4 EUR
APOSOR908142-1G APOSOR908142-5G
4-Hydroxy-N,N-dimethylbenzamide 98%
4-Hydroxy-N,N-dimethylbenzamide
Formel: C₉H₁₁NO₂
Molecular Weight: 165.189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01168919
CAS-Nummer: 20876-99-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR