Methyl 3-(acetyloxy)benzoate 98%

Lieferant: Apollo Scientific

OR908186-1G OR908186-5G
APOSOR908186-1GEA 248 EUR
APOSOR908186-1G APOSOR908186-5G
Methyl 3-(acetyloxy)benzoate 98%
Methyl 3-(acetyloxy)benzoate
Formel: C₁₀H₁₀O₄
Molecular Weight: 194.184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00092814
CAS-Nummer: 24781-23-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR