2-Morpholinopyridin-4-amine 98%

Lieferant: Apollo Scientific

OR908236-100MG OR908236-1G OR908236-250MG
APOSOR908236-100MGEA 78.3 EUR
APOSOR908236-100MG APOSOR908236-1G APOSOR908236-250MG
2-Morpholinopyridin-4-amine 98%
2-Morpholinopyridin-4-amine
Formel: C₉H₁₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD02945666
CAS-Nummer: 35980-77-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR