2-formyl-3-methoxyphenylboronic acid 95%
Lieferant: Apollo Scientific
Synonyme:
(2-Formyl-3-methoxyphenyl)boronic acid
OR908289-1G
OR908289-5G
APOSOR908289-1GEA
94.4
EUR
APOSOR908289-1G
APOSOR908289-5G
2-formyl-3-methoxyphenylboronic acid 95%
2-formyl-3-methoxyphenylboronic acid
Formel:
C₈H₉BO₄ Molecular Weight: 179,966 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319042 CAS-Nummer: 958030-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...