2-(Cyclopentyl)ethenyl-1-boronic acid 96%
Lieferant: Apollo Scientific
Synonyme:
(E)-(2-Cyclopentylvinyl)boronic acid
OR908337-1G
OR908337-250MG
APOSOR908337-1GEA
1290
EUR
APOSOR908337-1G
APOSOR908337-250MG
2-(Cyclopentyl)ethenyl-1-boronic acid 96%
2-(Cyclopentyl)ethenyl-1-boronic acid
Formel:
C₇H₁₃BO₂ Molecular Weight: 139.988 g/mol Schmelzpunkt: 70…72 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD09840294 CAS-Nummer: 161282-93-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...