3-Cyano-4-methoxyphenylboronic acid 95%

Lieferant: Apollo Scientific

OR908389-1G OR908389-5G
APOSOR908389-1GEA 202 EUR
APOSOR908389-1G APOSOR908389-5G
3-Cyano-4-methoxyphenylboronic acid 95%
3-Cyano-4-methoxyphenylboronic acid
Formel: C₈H₈BNO₃
Molecular Weight: 176,965 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10697415
CAS-Nummer: 911210-48-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR