1-Methyl-2-oxopyridine-3-boronic acid 95%

Lieferant: Apollo Scientific
Achtung

OR908489-1G OR908489-5G
APOSOR908489-1GEA 246 EUR
APOSOR908489-1G APOSOR908489-5G
1-Methyl-2-oxopyridine-3-boronic acid 95%
1-Methyl-2-oxopyridine-3-boronic acid
Formel: C₆H₈BNO₃
Molecular Weight: 152,944 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD23699496
CAS-Nummer: 1454558-46-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR