2-Isopropoxy-5-methylpyridine-3-boronic acid 98%

Lieferant: Apollo Scientific

OR908561-1G OR908561-5G
APOSOR908561-1GEA 219 EUR
APOSOR908561-1G APOSOR908561-5G
2-Isopropoxy-5-methylpyridine-3-boronic acid 98%
2-Isopropoxy-5-methylpyridine-3-boronic acid
Formel: C₉H₁₄BNO₃
Molecular Weight: 195,023 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19237189
CAS-Nummer: 2096337-70-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR