[1-(Hydroxymethyl)cycloprop-1-yl]acetonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(Cyanomethyl)-1-(hydroxymethyl)cyclopropane, 2-[1-(Hydroxymethyl)cycloprop-1-yl]ethanonitrile, 2-(1-(Hydroxymethyl)cyclopropyl)acetonitrile
OR9086-10G
OR9086-25G
OR9086-5G
APOSOR9086-10GEA
314
EUR
APOSOR9086-10G
APOSOR9086-25G
APOSOR9086-5G
[1-(Hydroxymethyl)cycloprop-1-yl]acetonitrile 95%
[1-(Hydroxymethyl)cycloprop-1-yl]acetonitrile
Formel:
C₆H₉NO Molecular Weight: 111,142 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08235273 CAS-Nummer: 152922-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...