N-(4-Chloro-3-methoxyphenyl)acetamide 98%
Lieferant: Apollo Scientific
OR908903-1G
OR908903-5G
APOSOR908903-1GEA
145
EUR
APOSOR908903-1G
APOSOR908903-5G
N-(4-Chloro-3-methoxyphenyl)acetamide 98%
N-(4-Chloro-3-methoxyphenyl)acetamide
Formel:
C₉H₁₀ClNO₂ Molecular Weight: 199,634 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11053828 CAS-Nummer: 98446-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...