Methyl 5-cyclopropylpyrazole-4-carboxylate 96%
Lieferant: Apollo Scientific
Synonyme:
Methyl 5-cyclopropyl-1H-pyrazole-4-carboxylate
OR909061-1G
OR909061-25G
OR909061-5G
APOSOR909061-1GEA
58
EUR
APOSOR909061-1G
APOSOR909061-25G
APOSOR909061-5G
Methyl 5-cyclopropylpyrazole-4-carboxylate 96%
Methyl 5-cyclopropylpyrazole-4-carboxylate
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166,177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11855888 CAS-Nummer: 1150164-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...